Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015822.2 | NP_056637.1 | 3 - 4 | 817 | 64 | |
GenBank mRNA | AF176521.1 | - | 3 - 4 | 524 | 64 | |
AK080863.1 | - | 3 - 4 | 826 | 64 | ||
AK142007.1 | - | 3 - 4 | 817 | 64 | ||
AK166128.1 | - | 3 - 4 | 698 | 65 | ||
AK167409.1 | - | 3 - 4 | 636 | 64 | ||
BC026835.1 | - | 2 - 3 | 483 | 64 | ||
BC037665.1 | - | 2 - 3 | 480 | 64 | ||
BC046330.1 | - | 3 - 4 | 531 | 64 | ||
BC067203.1 | - | 3 - 4 | 699 | 6334317 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |