Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183172.2 | NP_898995.1 | 8 - 9 | 1576 | 84 | |
GenBank mRNA | AY940666.1 | - | 8 - 9 | 1545 | 84 | |
BC051080.1 | - | 8 - 9 | 1573 | 6490136 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |