Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033314.3 | NP_201571.2 | 1 - 2 | 232 | 57 | |
GenBank mRNA | AF323958.1 | - | 1 - 2 | 101 | 57 | |
AK087350.1 | - | 1 - 2 | 235 | 57 | ||
AK166724.1 | - | 1 - 2 | 233 | 57 | ||
AK169706.1 | - | 1 - 2 | 235 | 57 | ||
BC035200.1 | - | 222 | 3158233 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |