Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000007399.mm.1 |
NM_011561.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007399.mm.1 |
NM_011561.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007399.mm.1 |
NM_172552.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007399.mm.1 |
NM_172552.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007399.mm.1 |
NR_037190.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007399.mm.1 |
TC1000000928.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000007398.mm.1 |
NM_011561.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007398.mm.1 |
NM_011561.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007398.mm.1 |
NM_172552.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007398.mm.1 |
NM_172552.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007398.mm.1 |
NR_037190.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000007398.mm.1 |
TC1000000928.mm.1 |
MTA TC |