Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081393.1 | NP_001074862.1 | 5 - 6 | 711 | 96 | |
GenBank mRNA | AK015498.1 | - | 5 - 6 | 715 | 96 | |
BC158097.1 | - | 4 - 5 | 700 | 9088178 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |