Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029961.2 | NP_084237.1 | 26 - 27 | 3519 | 80 | |
GenBank mRNA | AY766239.1 | - | 26 - 27 | 3483 | 80 | |
JQ655148.1 | - | 13 - 14 | 2998 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Mus musculus Probe spans exons |