Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlgn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172932.4 | NP_766520.2 | 3 - 4 | 826 | 80 | |
GenBank mRNA | AK039018.1 | - | 3 - 4 | 796 | 80 | |
BC069040.1 | - | 3 - 4 | 1205 | 80 | ||
BC150773.1 | - | 2 - 3 | 647 | 9087684 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |