Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172964.4 | NP_766552.3 | 5 - 6 | 827 | 93 | |
GenBank mRNA | AK087523.1 | - | 5 - 6 | 840 | 93 | |
AK133720.1 | - | 5 - 6 | 669 | 93 | ||
AK141901.1 | - | 5 - 6 | 777 | 93 | ||
AK173147.1 | - | 4 - 5 | 454 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |