Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290628.1 | NP_001277557.1 | 4 - 5 | 776 | 139 | |
NM_139061.5 | NP_620692.3 | 4 - 5 | 812 | 139 | ||
GenBank mRNA | AF424699.2 | - | 4 - 5 | 563 | 139 | |
AK077306.1 | - | 2 - 3 | 194 | 139 | ||
BC025012.2 | - | 4 - 5 | 487 | 5042879 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |