Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dzip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK013136.1 | - | 6 - 7 | 619 | 102 | |
AK032834.1 | - | 12 - 13 | 1228 | 102 | ||
AK129256.1 | - | 15 - 16 | 2029 | 102 | ||
AK141247.1 | - | 17 - 18 | 2445 | 102 | ||
BC051931.1 | - | 3 - 4 | 427 | 102 | ||
BC060196.1 | - | 12 - 13 | 2786 | 102 | ||
BC098211.1 | - | 17 - 18 | 2517 | 6825425 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |