Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dzip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK129256.1 | - | 3 - 4 | 746 | 56 | |
AK141247.1 | - | 5 - 6 | 1159 | 56 | ||
BC098211.1 | - | 5 - 6 | 1231 | 6825425 | 56 | |
BQ963394.1 | - | 3 - 4 | 732 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |