Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dzip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK013136.1 | - | 3 - 4 | 192 | 149 | |
AK032834.1 | - | 9 - 10 | 801 | 149 | ||
AK129256.1 | - | 12 - 13 | 1602 | 149 | ||
BC060196.1 | - | 9 - 10 | 2359 | 149 | ||
BC098211.1 | - | 14 - 15 | 2090 | 6825425 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |