Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029578.3 | NP_083854.3 | 1 - 2 | 124 | 95 | |
GenBank mRNA | AK011439.1 | - | 1 - 2 | 103 | 94 | |
AK011555.1 | - | 1 - 2 | 115 | 95 | ||
AK154341.1 | - | 1 - 2 | 101 | 95 | ||
AK165685.1 | - | 1 - 2 | 98 | 95 | ||
BC021419.1 | - | 1 - 2 | 100 | 5012877 | 95 | |
BY710811.1 | - | 1 - 2 | 112 | 95 | ||
CB953719.1 | - | 1 - 2 | 127 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |