Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144798.6 | NP_659047.2 | 4 - 5 | 263 | 94 | |
GenBank mRNA | AF233346.1 | - | 3 - 4 | 215 | 94 | |
AK082907.1 | - | 4 - 5 | 240 | 94 | ||
BC066162.1 | - | 4 - 5 | 230 | 30471027 | 94 | |
BC068169.1 | - | 4 - 5 | 252 | 30542277 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |