Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF411608.1 | - | 6 - 7 | 623 | 107 | |
BC012434.1 | - | 3 - 4 | 2408 | 107 | ||
BC071237.1 | - | 6 - 7 | 833 | 5003738 | 107 | |
BC106125.1 | - | 6 - 7 | 841 | 107 | ||
BC117075.1 | - | 6 - 7 | 689 | 40126394 | 107 | |
BC119092.1 | - | 6 - 7 | 689 | 8733841 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |