Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008549.2 | NP_032575.2 | 9 - 10 | 1664 | 151 | |
GenBank mRNA | AK144763.1 | - | 2579 | 151 | ||
BC138371.1 | - | 9 - 10 | 1649 | 8861391 | 151 | |
BC138372.1 | - | 9 - 10 | 1649 | 8861392 | 151 | |
X61172.1 | - | 9 - 10 | 1664 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |