Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172554.2 | NP_766142.2 | 6 - 7 | 645 | 120 | |
GenBank mRNA | AK038670.1 | - | 6 - 7 | 589 | 120 | |
AK038681.1 | - | 6 - 7 | 589 | 120 | ||
AK039575.1 | - | 6 - 7 | 646 | 120 | ||
AK046891.1 | - | 6 - 7 | 587 | 120 | ||
AK122406.1 | - | 6 - 7 | 719 | 120 | ||
AK134967.1 | - | 6 - 7 | 594 | 120 | ||
AK162922.1 | - | 6 - 7 | 583 | 120 | ||
BC058772.1 | - | 6 - 7 | 586 | 120 | ||
BC080664.1 | - | 6 - 7 | 584 | 120 | ||
BC117788.1 | - | 6 - 7 | 587 | 40092137 | 120 | |
BC117789.2 | - | 6 - 7 | 587 | 40092140 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |