Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027892.2 | NP_082168.1 | 10 - 11 | 1447 | 85 | |
GenBank mRNA | AK004785.1 | - | 10 - 11 | 1447 | 85 | |
AK030460.1 | - | 1 - 2 | 49 | 85 | ||
AK035230.1 | - | 2 - 3 | 234 | 85 | ||
AK139570.1 | - | 10 - 11 | 1333 | 85 | ||
BC125381.1 | - | 11 - 12 | 1516 | 40129896 | 85 | |
BC137630.1 | - | 11 - 12 | 1516 | 8860649 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |