Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144844.2 | NP_659093.2 | 15 - 16 | 1380 | 81 | |
GenBank mRNA | AK148002.1 | - | 15 - 16 | 1370 | 81 | |
AY046947.1 | - | 15 - 16 | 1397 | 81 | ||
BC006915.1 | - | 15 - 16 | 1377 | 3601148 | 81 | |
BC049802.1 | - | 15 - 16 | 1346 | 6332730 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |