Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008446.2 | NP_032472.2 | 11 - 12 | 1737 | 96 | |
GenBank mRNA | AK084837.1 | - | 11 - 12 | 1714 | 96 | |
AK135179.1 | - | 11 - 12 | 1685 | 96 | ||
AK135430.1 | - | 11 - 12 | 1231 | 96 | ||
AK171938.1 | - | 11 - 12 | 1648 | 96 | ||
BC050946.1 | - | 11 - 12 | 1675 | 6822283 | 96 | |
BC054537.1 | - | 11 - 12 | 1659 | 96 | ||
D12646.1 | - | 11 - 12 | 2064 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |