Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198303.2 | NP_938045.2 | 1 - 2 | 317 | 85 | |
GenBank mRNA | AK145146.1 | - | 1 - 2 | 204 | 85 | |
AK145732.1 | - | 1 - 2 | 318 | 85 | ||
AK163527.1 | - | 1 - 2 | 221 | 85 | ||
BC018347.1 | - | 1 - 2 | 173 | 85 | ||
BC037150.1 | - | 1 - 2 | 96 | 85 | ||
BC040746.1 | - | 1 - 2 | 188 | 85 | ||
BC060288.1 | - | 1 - 2 | 157 | 85 | ||
BC103806.1 | - | 1 - 2 | 198 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |