Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sycp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011517.2 | NP_035647.2 | 7 - 8 | 689 | 145 | |
GenBank mRNA | AK131949.1 | - | 7 - 8 | 613 | 145 | |
BC099552.1 | - | 6 - 7 | 494 | 145 | ||
BC132079.1 | - | 7 - 8 | 690 | 40130356 | 145 | |
BC138509.1 | - | 7 - 8 | 690 | 8861529 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |