Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128180.1 | NP_001121652.1 | 6 - 7 | 1251 | 76 | |
NM_001128181.1 | NP_001121653.1 | 7 - 8 | 1253 | 76 | ||
NM_027712.3 | NP_081988.3 | 7 - 8 | 2238 | 76 | ||
NM_177639.6 | NP_808307.2 | 8 - 9 | 2268 | 76 | ||
GenBank mRNA | AK034182.1 | - | 2 - 3 | 127 | 76 | |
AK035763.1 | - | 11 - 12 | 2819 | 76 | ||
AK136926.1 | - | 7 - 8 | 1214 | 76 | ||
AK147448.1 | - | 11 - 12 | 3055 | 76 | ||
AK161941.1 | - | 6 - 7 | 1226 | 76 | ||
AK163689.1 | - | 6 - 7 | 1232 | 76 | ||
AK220515.1 | - | 10 - 11 | 3118 | 76 | ||
AY243846.2 | - | 7 - 8 | 2019 | 76 | ||
BC062120.1 | - | 8 - 9 | 2269 | 6822047 | 76 | |
BC094369.1 | - | 6 - 7 | 1447 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |