Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175930.5 | NP_787126.3 | 9 - 10 | 1231 | 68 | |
GenBank mRNA | AK029995.1 | - | 9 - 10 | 1232 | 68 | |
AK030016.1 | - | 9 - 10 | 1185 | 68 | ||
AK162220.1 | - | 7 - 8 | 864 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |