Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008073.3 | NP_032099.1 | 9 - 10 | 1540 | 64 | |
GenBank mRNA | AF233775.1 | - | 9 - 10 | 1156 | 64 | |
AF233776.1 | - | 9 - 10 | 1156 | 64 | ||
AF233777.1 | - | 9 - 10 | 1156 | 64 | ||
AF233778.1 | - | 9 - 10 | 1156 | 64 | ||
AF233779.1 | - | 9 - 10 | 1156 | 64 | ||
AF233780.1 | - | 9 - 10 | 1156 | 64 | ||
AF233781.1 | - | 9 - 10 | 1156 | 64 | ||
AF233782.1 | - | 9 - 10 | 1156 | 64 | ||
AF233783.1 | - | 9 - 10 | 1156 | 64 | ||
AF233784.1 | - | 9 - 10 | 1156 | 64 | ||
AF233785.1 | - | 9 - 10 | 1156 | 64 | ||
AF233786.1 | - | 9 - 10 | 1156 | 64 | ||
AF233787.1 | - | 9 - 10 | 1156 | 64 | ||
AF233788.1 | - | 9 - 10 | 1156 | 64 | ||
AF233789.1 | - | 9 - 10 | 1156 | 64 | ||
AF233790.1 | - | 9 - 10 | 1156 | 64 | ||
AF233791.1 | - | 9 - 10 | 1156 | 64 | ||
AF233792.1 | - | 9 - 10 | 1156 | 64 | ||
AF233793.1 | - | 9 - 10 | 1156 | 64 | ||
AF233794.1 | - | 9 - 10 | 1156 | 64 | ||
AF233795.1 | - | 9 - 10 | 1156 | 64 | ||
AF233796.1 | - | 9 - 10 | 1156 | 64 | ||
AF233797.1 | - | 9 - 10 | 1156 | 64 | ||
AF233798.1 | - | 9 - 10 | 1156 | 64 | ||
AF233799.1 | - | 9 - 10 | 1156 | 64 | ||
AF233800.1 | - | 9 - 10 | 1156 | 64 | ||
AF233801.1 | - | 9 - 10 | 1156 | 64 | ||
AF233802.1 | - | 9 - 10 | 1156 | 64 | ||
AK047085.1 | - | 9 - 10 | 1118 | 64 | ||
BC031762.1 | - | 9 - 10 | 1358 | 4504205 | 64 | |
M62374.1 | - | 9 - 10 | 1503 | 64 | ||
X56313.1 | - | 2 - 3 | 232 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |