Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011546.3 | NP_035676.1 | 4 - 5 | 670 | 86 | |
GenBank mRNA | AK140038.1 | - | 4 - 5 | 832 | 86 | |
BC090987.1 | - | 4 - 5 | 635 | 86 | ||
BC139768.1 | - | 4 - 5 | 639 | 40111224 | 86 | |
BC139769.1 | - | 4 - 5 | 639 | 40111227 | 86 | |
D76432.1 | - | 4 - 5 | 639 | 86 | ||
U26259.1 | - | 4 - 5 | 656 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |