Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013814.1 | NP_001013836.1 | 1 - 2 | 98 | 112 | |
GenBank mRNA | BC132277.1 | - | 1 - 2 | 150 | 40130554 | 112 |
BC132279.1 | - | 1 - 2 | 150 | 40130556 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |