Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016802.4 | NP_058082.2 | 5 - 6 | 833 | 61 | |
GenBank mRNA | AF014371.1 | - | 4 - 5 | 541 | 61 | |
AF178958.1 | - | 4 - 5 | 428 | 61 | ||
AF178959.1 | - | 4 - 5 | 428 | 61 | ||
AF178960.1 | - | 4 - 5 | 428 | 61 | ||
AF178961.1 | - | 4 - 5 | 428 | 61 | ||
AK077606.1 | - | 5 - 6 | 641 | 61 | ||
AK083624.1 | - | 5 - 6 | 640 | 61 | ||
AK150294.1 | - | 5 - 6 | 678 | 60 | ||
AK150475.1 | - | 5 - 6 | 684 | 61 | ||
AK152629.1 | - | 5 - 6 | 620 | 61 | ||
AK165512.1 | - | 5 - 6 | 616 | 61 | ||
AK172051.1 | - | 5 - 6 | 641 | 61 | ||
BC068115.1 | - | 5 - 6 | 833 | 6809149 | 61 | |
BC096423.1 | - | 5 - 6 | 619 | 3590185 | 61 | |
JN971019.1 | - | 3 - 4 | 407 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |