Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2w | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025773.3 | NP_080049.2 | 3 - 4 | 300 | 139 | |
GenBank mRNA | AK015348.1 | - | 3 - 4 | 254 | 139 | |
AK042870.1 | - | 3 - 4 | 250 | 139 | ||
AK044463.1 | - | 3 - 4 | 252 | 139 | ||
AK046086.1 | - | 3 - 4 | 254 | 139 | ||
AK050679.1 | - | 3 - 4 | 252 | 139 | ||
AK077997.1 | - | 3 - 4 | 225 | 139 | ||
AK082233.1 | - | 3 - 4 | 249 | 139 | ||
BC020124.1 | - | 3 - 4 | 252 | 4012785 | 139 | |
BY728312.1 | - | 3 - 4 | 300 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |