Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111274.1 | NP_001104744.1 | 4 - 5 | 658 | 81 | |
NM_010512.4 | NP_034642.2 | 4 - 5 | 740 | 81 | ||
GenBank mRNA | AY878193.1 | - | 4 - 5 | 538 | 81 | |
BC012409.1 | - | 3 - 4 | 395 | 4194295 | 81 | |
X04482.1 | - | 4 - 5 | 440 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |