Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plscr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195084.1 | NP_001182013.1 | 3 - 4 | 498 | 116 | |
NM_008880.3 | NP_032906.2 | 2 - 3 | 493 | 116 | ||
GenBank mRNA | AF015790.1 | - | 3 - 4 | 264 | 116 | |
AK002410.1 | - | 2 - 3 | 380 | 116 | ||
BC002012.1 | - | 3 - 4 | 364 | 3490475 | 116 | |
BC011299.1 | - | 2 - 3 | 375 | 3595010 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |