Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glipr1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027018.1 | NP_081294.1 | 3 - 4 | 669 | 74 | |
GenBank mRNA | AK005860.1 | - | 3 - 4 | 670 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Protease Mus musculus Probe spans exons |