Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148930.3 | NP_683732.1 | 11 - 12 | 1119 | 101 | |
GenBank mRNA | AJ309168.1 | - | 10 - 11 | 954 | 101 | |
AK134183.1 | - | 11 - 12 | 1114 | 101 | ||
AK146880.1 | - | 11 - 12 | 1114 | 101 | ||
BC003988.1 | - | 1 - 2 | 71 | 101 | ||
BC005719.1 | - | 6 - 7 | 724 | 101 | ||
BC019766.1 | - | 11 - 12 | 1064 | 101 | ||
BC023854.1 | - | 11 - 12 | 1104 | 5346780 | 101 | |
BC031899.1 | - | 11 - 12 | 1104 | 5362121 | 101 | |
BC042419.1 | - | 9 - 10 | 4355 | 101 | ||
BC043058.1 | - | 11 - 12 | 1056 | 6419695 | 101 | |
BC054729.1 | - | 11 - 12 | 1107 | 6419758 | 101 | |
BC054820.1 | - | 9 - 10 | 3833 | 101 | ||
BC060985.1 | - | 10 - 11 | 1023 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |