Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apeh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146226.2 | NP_666338.2 | 1 - 2 | 42 | 83 | |
GenBank mRNA | AK041271.1 | - | 1 - 2 | 39 | 83 | |
AK149671.1 | - | 1 - 2 | 44 | 83 | ||
AK149713.1 | - | 1 - 2 | 44 | 83 | ||
BC005716.1 | - | 1 - 2 | 28 | 83 | ||
CJ134183.1 | - | 1 - 2 | 33 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |