Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011251.3 | NP_035381.2 | 14 - 15 | 2669 | 126 | |
NM_029169.3 | NP_083445.1 | 14 - 15 | 2940 | 126 | ||
GenBank mRNA | AJ006486.2 | - | 14 - 15 | 2548 | 126 | |
AK013915.1 | - | 5 - 6 | 362 | 126 | ||
AK032150.1 | - | 9 - 10 | 921 | 126 | ||
AK077899.1 | - | 14 - 15 | 2847 | 126 | ||
AK145612.1 | - | 14 - 15 | 2829 | 126 | ||
AK164432.1 | - | 9 - 10 | 1026 | 126 | ||
AK220446.1 | - | 14 - 15 | 2818 | 126 | ||
BC026129.1 | - | 14 - 15 | 2694 | 3982861 | 126 | |
BC026130.1 | - | 14 - 15 | 2843 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |