Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146092.2 | NP_666204.2 | 8 - 9 | 776 | 63 | |
GenBank mRNA | AK144881.1 | - | 8 - 9 | 743 | 63 | |
BC020299.1 | - | 8 - 9 | 749 | 63 | ||
BC028647.1 | - | 8 - 9 | 723 | 1244243 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |