Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276704.1 | NP_001263633.1 | 9 - 10 | 1016 | 83 | |
NM_016813.2 | NP_058093.2 | 9 - 10 | 1030 | 83 | ||
GenBank mRNA | AF093140.1 | - | 9 - 10 | 1006 | 83 | |
AK077644.1 | - | 9 - 10 | 987 | 83 | ||
AK079583.1 | - | 9 - 10 | 1019 | 83 | ||
AK086527.1 | - | 9 - 10 | 1027 | 83 | ||
AK088634.1 | - | 9 - 10 | 987 | 83 | ||
AK133883.1 | - | 9 - 10 | 1019 | 83 | ||
AK143640.1 | - | 1017 | 83 | |||
AK167518.1 | - | 9 - 10 | 1018 | 83 | ||
BC005594.1 | - | 9 - 10 | 1030 | 3710546 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |