Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164634.1 | NP_001158106.1 | 7 - 8 | 1226 | 63 | |
NM_001164635.1 | NP_001158107.1 | 7 - 8 | 1134 | 63 | ||
NM_031194.5 | NP_112471.3 | 8 - 9 | 1306 | 63 | ||
GenBank mRNA | AB079895.1 | - | 6 - 7 | 963 | 63 | |
AF078869.1 | - | 7 - 8 | 1081 | 63 | ||
AK031962.1 | - | 8 - 9 | 1272 | 63 | ||
AK044336.1 | - | 7 - 8 | 1103 | 63 | ||
AK157891.1 | - | 7 - 8 | 1135 | 63 | ||
BC014762.1 | - | 7 - 8 | 1112 | 4239544 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |