Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193271.1 | NP_001180200.1 | 8 - 9 | 1624 | 79 | |
NM_010789.3 | NP_034919.1 | 8 - 9 | 1624 | 79 | ||
GenBank mRNA | AB221660.1 | - | 7 - 8 | 887 | 79 | |
AK132298.1 | - | 8 - 9 | 1404 | 79 | ||
AK134321.1 | - | 8 - 9 | 1619 | 79 | ||
AK140748.1 | - | 8 - 9 | 1628 | 79 | ||
AK146680.1 | - | 4 - 5 | 432 | 79 | ||
AK168684.1 | - | 8 - 9 | 1616 | 79 | ||
BC023689.1 | - | 8 - 9 | 1620 | 5344307 | 79 | |
U33629.1 | - | 8 - 9 | 1335 | 79 | ||
U33630.1 | - | 8 - 9 | 1335 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |