Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190401.2 | NP_001177330.2 | 16 - 17 | 2688 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |