Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194334.2 | NP_919315.2 | 9 - 10 | 2340 | 93 | |
GenBank mRNA | AK089175.1 | - | 9 - 10 | 2306 | 93 | |
AK129273.1 | - | 9 - 10 | 2326 | 93 | ||
AK156788.1 | - | 9 - 10 | 2327 | 93 | ||
BC056467.1 | - | 2 - 3 | 651 | 93 | ||
BC108420.1 | - | 4 - 5 | 945 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |