Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194334.2 | NP_919315.2 | 3 - 4 | 732 | 82 | |
GenBank mRNA | AK089175.1 | - | 3 - 4 | 697 | 82 | |
AK129273.1 | - | 3 - 4 | 718 | 82 | ||
AK156788.1 | - | 3 - 4 | 719 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |