Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900023406.mm.1 |
NM_001039147.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023406.mm.1 |
NM_001039147.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023406.mm.1 |
NM_024431.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023406.mm.1 |
NM_024431.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023406.mm.1 |
TC0900002820.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023410.mm.1 |
NM_001039147.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023410.mm.1 |
NM_001039147.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023410.mm.1 |
TC0900002820.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023408.mm.1 |
NM_001039147.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023408.mm.1 |
NM_001039147.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023408.mm.1 |
NM_024431.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023408.mm.1 |
NM_024431.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023408.mm.1 |
TC0900002820.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023409.mm.1 |
NM_001039147.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023409.mm.1 |
NM_001039147.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023409.mm.1 |
NM_024431.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023409.mm.1 |
NM_024431.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023409.mm.1 |
TC0900002820.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023400.mm.1 |
NM_001039147.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023400.mm.1 |
NM_001039147.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023400.mm.1 |
NM_024431.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023400.mm.1 |
NM_024431.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023400.mm.1 |
TC0900002820.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023403.mm.1 |
NM_001039147.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023403.mm.1 |
NM_001039147.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023403.mm.1 |
NM_024431.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023403.mm.1 |
NM_024431.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023403.mm.1 |
TC0900002820.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023412.mm.1 |
NM_001039147.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023412.mm.1 |
NM_001039147.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023412.mm.1 |
NM_024431.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023412.mm.1 |
NM_024431.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023412.mm.1 |
TC0900002820.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023405.mm.1 |
NM_001039147.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023405.mm.1 |
NM_001039147.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023405.mm.1 |
NM_024431.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023405.mm.1 |
NM_024431.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023405.mm.1 |
TC0900002820.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023407.mm.1 |
NM_001039147.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023407.mm.1 |
NM_001039147.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023407.mm.1 |
NM_024431.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023407.mm.1 |
NM_024431.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023407.mm.1 |
TC0900002820.mm.1 |
MTA TC |