Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144868.3 | NP_659117.2 | 8 - 9 | 2768 | 90 | |
GenBank mRNA | AF237953.1 | - | 6 - 7 | 785 | 90 | |
AK171516.1 | - | 7 - 8 | 3005 | 90 | ||
BC025838.1 | - | 2 - 3 | 301 | 90 | ||
BC038056.1 | - | 3 - 4 | 876 | 90 | ||
BC054751.1 | - | 8 - 9 | 2047 | 90 | ||
BC055289.1 | - | 9 - 10 | 2571 | 90 | ||
BC096538.1 | - | 8 - 9 | 2768 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |