Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024560.2 | NP_001019731.1 | 10 - 11 | 1029 | 74 | |
GenBank mRNA | AK037118.1 | - | 1603 | 74 | ||
AK047234.1 | - | 10 - 11 | 1005 | 74 | ||
AK143441.1 | - | 8 - 9 | 1724 | 74 | ||
BC048922.1 | - | 10 - 11 | 1029 | 6311864 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |