Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199247.1 | NP_001186176.1 | 9 - 10 | 984 | 74 | |
NM_001199248.1 | NP_001186177.1 | 8 - 9 | 1083 | 74 | ||
NM_001199249.1 | NP_001186178.1 | 8 - 9 | 1083 | 74 | ||
NM_178637.2 | NP_848752.1 | 10 - 11 | 1140 | 74 | ||
NR_037603.1 | 7 - 8 | 1016 | 74 | |||
GenBank mRNA | AB055409.1 | - | 7 - 8 | 994 | 74 | |
AF528194.1 | - | 6 - 7 | 694 | 74 | ||
AF528195.1 | - | 9 - 10 | 1075 | 74 | ||
AF528196.1 | - | 8 - 9 | 919 | 74 | ||
AK143674.1 | - | 8 - 9 | 1035 | 74 | ||
AK146352.1 | - | 10 - 11 | 1097 | 74 | ||
AK205910.1 | - | 1 - 2 | 59 | 74 | ||
BC022768.1 | - | 1 - 2 | 65 | 74 | ||
BC110675.1 | - | 10 - 11 | 1081 | 4011689 | 74 | |
BC129967.1 | - | 8 - 9 | 918 | 74 | ||
BC129968.1 | - | 8 - 9 | 919 | 40106268 | 74 | |
CA317697.1 | - | 3 - 4 | 378 | 74 | ||
DQ139980.1 | - | 9 - 10 | 1075 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |