Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199247.1 | NP_001186176.1 | 2 - 3 | 145 | 70 | |
NM_001199248.1 | NP_001186177.1 | 1 - 2 | 244 | 70 | ||
NM_001199249.1 | NP_001186178.1 | 1 - 2 | 244 | 70 | ||
NM_178637.2 | NP_848752.1 | 2 - 3 | 145 | 70 | ||
GenBank mRNA | AF528195.1 | - | 1 - 2 | 80 | 70 | |
AF528196.1 | - | 1 - 2 | 80 | 70 | ||
AK143674.1 | - | 1 - 2 | 196 | 70 | ||
AK146352.1 | - | 2 - 3 | 102 | 70 | ||
AK159568.1 | - | 1 - 2 | 177 | 70 | ||
AK160077.1 | - | 1 - 2 | 177 | 70 | ||
AK195015.1 | - | 1 - 2 | 47 | 70 | ||
BC110675.1 | - | 2 - 3 | 86 | 4011689 | 70 | |
BC129967.1 | - | 1 - 2 | 79 | 70 | ||
BC129968.1 | - | 1 - 2 | 80 | 40106268 | 70 | |
DQ139980.1 | - | 1 - 2 | 80 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |