Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK078127.1 | - | 1 - 2 | 72 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |