Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026763.2 | NP_081039.2 | 22 - 23 | 4853 | 91 | |
GenBank mRNA | AB370265.1 | - | 22 - 23 | 4945 | 91 | |
AM231153.1 | - | 16 - 17 | 2233 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |