Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB370265.1 | - | 28 - 29 | 5286 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |